missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Phenyl Ketone 98.0+%, TCI America™

Catalog No. C134525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C134525G 25 g
C1345500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C134525G Supplier TCI America Supplier No. C134525G

Chemical Identifiers

CAS 712-50-5
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00001467
InChI Key BMFYCFSWWDXEPB-UHFFFAOYSA-N
Synonym cyclohexyl phenyl ketone, cyclohexyl phenyl methanone, benzoylcyclohexane, cyclohexylphenylketone, benzoyl cyclohexane, phenyl cyclohexyl ketone, methanone, cyclohexylphenyl, cyclohexyl-phenyl-methanone, usaf kf-3, cyclohexylphenyl ketone
PubChem CID 12837
IUPAC Name cyclohexyl(phenyl)methanone
SMILES C1CCC(CC1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 168°C
Quantity 25 g
Formula Weight 188.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexyl Phenyl Ketone
EINECSNumber (9)-2012
RTECSNumber OB1760000
TSCA Yes