accessibility menu, dialog, popup

UserName

CAS RN 712-50-5

IUPAC Name: cyclohexyl(phenyl)methanone
Synonyms: cyclohexylphenylketone
Molecular Weight (g/mol): 188.27
Molecular Formula: C13H16O
InChi Key: BMFYCFSWWDXEPB-UHFFFAOYSA-N
SMILES: O=C(C1CCCCC1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)