missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl phenyl ketone, 98%

Catalog No. AAB2481606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2481606 5 g
AAB2481614 25 g
AAB2481622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2481606 Supplier Thermo Scientific Chemicals Supplier No. B2481606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 712-50-5 | C13H16O | 188.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 712-50-5
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00001467
InChI Key BMFYCFSWWDXEPB-UHFFFAOYSA-N
Synonym cyclohexyl phenyl ketone, cyclohexyl phenyl methanone, benzoylcyclohexane, cyclohexylphenylketone, benzoyl cyclohexane, phenyl cyclohexyl ketone, methanone, cyclohexylphenyl, cyclohexyl-phenyl-methanone, usaf kf-3, cyclohexylphenyl ketone
PubChem CID 12837
IUPAC Name cyclohexyl(phenyl)methanone
SMILES C1CCC(CC1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 52°C to 57°C
Boiling Point 165°C to 168°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2046712
Formula Weight 188.27
Percent Purity 98%
Chemical Name or Material Cyclohexyl phenyl ketone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-923-5
RTECSNumber OB1760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only