missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexyl p-Toluenesulfonate 98.0+%, TCI America™
Supplier: TCI America C236325G
Specifications
| Cyclohexyl p-Toluenesulfonate | |
| 45°C | |
| C13H18O3S | |
| 25 g | |
| OHHPZPDQZMUTCA-UHFFFAOYSA-N | |
| cyclohexyl 4-methylbenzenesulfonate | |
| 13722 | |
| ≥98.0% (HPLC) |
| 953-91-3 | |
| White-Yellow | |
| MFCD00014291 | |
| cyclohexyl p-toluenesulfonate, cyclohexyl tosylate, cyclohexyl toluenesulfonate, cyclohexylp-toluenesulfonate, cyclohexyl p-toluenesulphonate, benzenesulfonic acid, 4-methyl-, cyclohexyl ester, p-toluenesulfonic acid, cyclohexyl ester, cyclohexyl-p-toluene sulfonate, benzenesulfonic acid, cyclohexyl ester, acmc-20aj06 | |
| CC1=CC=C(C=C1)S(=O)(=O)OC2CCCCC2 | |
| 254.344 | |
| 254.34 | |
| Crystalline Powder |
Chemical Identifiers
| 953-91-3 | |
| 254.344 | |
| OHHPZPDQZMUTCA-UHFFFAOYSA-N | |
| 13722 | |
| CC1=CC=C(C=C1)S(=O)(=O)OC2CCCCC2 |
| C13H18O3S | |
| MFCD00014291 | |
| cyclohexyl p-toluenesulfonate, cyclohexyl tosylate, cyclohexyl toluenesulfonate, cyclohexylp-toluenesulfonate, cyclohexyl p-toluenesulphonate, benzenesulfonic acid, 4-methyl-, cyclohexyl ester, p-toluenesulfonic acid, cyclohexyl ester, cyclohexyl-p-toluene sulfonate, benzenesulfonic acid, cyclohexyl ester, acmc-20aj06 | |
| cyclohexyl 4-methylbenzenesulfonate |
Safety and Handling
TSCA : Yes