missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methyl methylphosphonate, 99%, Thermo Scientific Chemicals

Catalog No. p-7059332
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4442602 0.5 g
AA4442604 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4442602 Supplier Thermo Scientific Chemicals Supplier No. 04442602
Only null left

Cyclohexyl methyl methylphosphonate or phosphonic acid is used as a micro-nano electronic materials have extended self-assembled monolayers (SAMs) beyond conventional gold/thiol systems. To expand the choice of the substrate used for the preparation of SAMs, the chemical functionalities in film-forming molecules can be altered by introducing phosphate or phosphonate groups. Such polar acidic molecules are capable of interacting with diverse metal-oxide surfaces (e.g., Al2O3, Ta2O5, NbO5, ZrO2 and TiO2) and form films with a similar degree of ordering as for alkyl thiol SAMs on gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7040-52-0
Molecular Formula C8H17O3P
Molecular Weight (g/mol) 192.195
MDL Number MFCD08064211
InChI Key UXITUBFQUAHYSL-UHFFFAOYSA-N
Synonym cyclohexyl methyl methylphosphonate, phosphonic acid, methyl-, cyclohexyl methyl ester, cyclohexyl methyl methyl-phosphonate, methoxy methyl phosphoryl oxycyclohexane, methylphosphonic acid methylcyclohexyl ester
PubChem CID 574009
IUPAC Name [methoxy(methyl)phosphoryl]oxycyclohexane
SMILES COP(=O)(C)OC1CCCCC1

Specifications

Quantity 0.5 g
Solubility Information It is sparingly soluble in both organic solvents and water,but become more soluble in water when neutralized to phosphonates at neutral to high pH.
Formula Weight 192.09
Percent Purity 99%
Chemical Name or Material Cyclohexyl methyl methylphosphonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only