accessibility menu, dialog, popup

UserName

CAS RN 7040-52-0

IUPAC Name: cyclohexyl methyl methylphosphonate
Synonyms: cyclohexyl methyl methylphosphonate
Molecular Weight (g/mol): 192.2
Molecular Formula: C8H17O3P
InChi Key: UXITUBFQUAHYSL-UHFFFAOYNA-N
SMILES: COP(C)(=O)OC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results