missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methyl ketone, 95%

Catalog No. AAB2100006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2100006 5 g
AAB2100014 25 g
AAB2100022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2100006 Supplier Thermo Scientific Chemicals Supplier No. B2100006
Only null left

CAS: 823-76-7 | C8H14O | 126.199 g/mol

It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 823-76-7
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00040418
InChI Key RIFKADJTWUGDOV-UHFFFAOYSA-N
Synonym 1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl
PubChem CID 13207
IUPAC Name 1-cyclohexylethanone
SMILES CC(=O)C1CCCCC1

Specifications

Melting Point -34°C
Density 0.917
Boiling Point 181°C to 183°C
Flash Point 66°C (151°F)
Assay Percent Range 0.95
Refractive Index 1.452
Quantity 5 g
Beilstein 507229
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 126.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Cyclohexyl methyl ketone
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 212-517-0
RTECSNumber KM5638850
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only