missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I018710ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I018710ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I018710ML Supplier TCI America Supplier No. I018710ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1122-82-3
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.232
MDL Number MFCD00003841
InChI Key MZSJGCPBOVTKHR-UHFFFAOYSA-N
Synonym cyclohexyl isothiocyanate, cyclohexane, isothiocyanato, isothiocyanocyclohexane, cyclohexyl-isothiocyanat, isothiocyanic acid, cyclohexyl ester, cyclohexylisothiocyanate, cyclohexyl isothiocyanate, isothiocyanato, cyclohexyl-isothiocyanat german, isothiocyanato-cyclohexane, isothiocyanic acid cyclohexyl ester
PubChem CID 14289
IUPAC Name isothiocyanatocyclohexane
SMILES C1CCC(CC1)N=C=S

Specifications

Color Yellow
Boiling Point 219°C
Quantity 10 mL
UN Number 1760
Formula Weight 141.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl Isothiocyanate
RTECSNumber NX8480000
TSCA Yes