missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Isocyanide 98.0+%, TCI America™

Catalog No. p-7124981
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14381G 1 g
C14385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14381G Supplier TCI America Supplier No. C14381G
Only null left

Chemical Identifiers

CAS 931-53-3
Molecular Formula C7H11N
Molecular Weight (g/mol) 109.172
MDL Number MFCD00003839
InChI Key XYZMOVWWVXBHDP-UHFFFAOYSA-N
Synonym cyclohexyl isocyanide, cyclohexane, isocyano, cyclohexylisocyanide, cyclohexaneisonitrile, cyclohexyl isonitrile, n-cyclohexylisocyanide, cyi, cyclohexylisonitrile, isocyano-cyclohexane, cyclohexyliminocarbene
PubChem CID 79129
ChEBI CHEBI:17966
IUPAC Name isocyanocyclohexane
SMILES [C-]#[N+]C1CCCCC1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 1 g
UN Number 2810
Formula Weight 109.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl Isocyanide