accessibility menu, dialog, popup

UserName

CAS RN 931-53-3

IUPAC Name: isocyanocyclohexane
Synonyms: cyclohexyl isocyanide
Molecular Weight (g/mol): 109.17
Molecular Formula: C7H11N
InChi Key: XYZMOVWWVXBHDP-UHFFFAOYSA-N
SMILES: [C-]#[N+]C1CCCCC1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)