missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl ethyl ketone, 99%

CAS: 1123-86-0 | C9H16O | 140.226 g/mol

$430.74 - $1694.70

Chemical Identifiers

CAS 1123-86-0
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00060814
InChI Key AMHOPTNGSNYSBL-UHFFFAOYSA-N
Synonym ethyl cyclohexyl ketone, cyclohexylethylketone, 1-propanone, 1-cyclohexyl, cyclohexyl ethyl ketone, ketone, cyclohexyl ethyl, 1-cyclohexyl-1-propanone, 1-cyclohexyl-propan-1-one, propanoylcyclohexane, 1-methylacetylcyclohexan, acmc-1c9ub
PubChem CID 70748
IUPAC Name 1-cyclohexylpropan-1-one
SMILES CCC(=O)C1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2040906
View Documents
Thermo Scientific Chemicals
B2040906
5 g
Each for $430.74
Only null left
 
AAB2040914
View Documents
Thermo Scientific Chemicals
B2040914
25 g
Each for $1,694.70
Only null left
 
Description

Description

Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

Solubility
Soluble in organic solvents.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

1123-86-0
140.226
AMHOPTNGSNYSBL-UHFFFAOYSA-N
70748
CCC(=O)C1CCCCC1
C9H16O
MFCD00060814
ethyl cyclohexyl ketone, cyclohexylethylketone, 1-propanone, 1-cyclohexyl, cyclohexyl ethyl ketone, ketone, cyclohexyl ethyl, 1-cyclohexyl-1-propanone, 1-cyclohexyl-propan-1-one, propanoylcyclohexane, 1-methylacetylcyclohexan, acmc-1c9ub
1-cyclohexylpropan-1-one

Specifications

1123-86-0
C9H16O
5 g
Soluble in organic solvents.
CCC(=O)C1CCCCC1
140.226
140.23
Cyclohexyl ethyl ketone
0.91
MFCD00060814
ethyl cyclohexyl ketone, cyclohexylethylketone, 1-propanone, 1-cyclohexyl, cyclohexyl ethyl ketone, ketone, cyclohexyl ethyl, 1-cyclohexyl-1-propanone, 1-cyclohexyl-propan-1-one, propanoylcyclohexane, 1-methylacetylcyclohexan, acmc-1c9ub
AMHOPTNGSNYSBL-UHFFFAOYSA-N
1-cyclohexylpropan-1-one
70748
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-380-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only