CAS RN 1123-86-0
IUPAC Name:
1-cyclohexylpropan-1-one
Synonyms:
1-cyclohexyl-propan-1-one
Molecular Weight (g/mol):
140.23
Molecular Formula:
C9H16O
InChi Key:
AMHOPTNGSNYSBL-UHFFFAOYSA-N
SMILES:
CCC(=O)C1CCCCC1
1
–
1
of
1
results
1
Promotions Available
Cyclohexyl ethyl ketone, 99%
Cyclohexyl ethyl ketone, 99%
CAS: 1123-86-0 Molecular Formula: C9H16O Molecular Weight (g/mol): 140.226 MDL Number: MFCD00060814 InChI Key: AMHOPTNGSNYSBL-UHFFFAOYSA-N Synonym: ethyl cyclohexyl ketone,cyclohexylethylketone,1-propanone, 1-cyclohexyl,cyclohexyl ethyl ketone,ketone, cyclohexyl ethyl,1-cyclohexyl-1-propanone,1-cyclohexyl-propan-1-one,propanoylcyclohexane,1-methylacetylcyclohexan,acmc-1c9ub PubChem CID: 70748 IUPAC Name: 1-cyclohexylpropan-1-one SMILES: CCC(=O)C1CCCCC1