missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl ethyl ketone, 99%

Catalog No. AAB2040906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2040906 5 g
AAB2040914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2040906 Supplier Thermo Scientific Chemicals Supplier No. B2040906

CAS: 1123-86-0 | C9H16O | 140.226 g/mol

Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

Solubility
Soluble in organic solvents.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.

Chemical Identifiers

CAS 1123-86-0
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00060814
InChI Key AMHOPTNGSNYSBL-UHFFFAOYSA-N
Synonym ethyl cyclohexyl ketone, cyclohexylethylketone, 1-propanone, 1-cyclohexyl, cyclohexyl ethyl ketone, ketone, cyclohexyl ethyl, 1-cyclohexyl-1-propanone, 1-cyclohexyl-propan-1-one, propanoylcyclohexane, 1-methylacetylcyclohexan, acmc-1c9ub
PubChem CID 70748
IUPAC Name 1-cyclohexylpropan-1-one
SMILES CCC(=O)C1CCCCC1

Specifications

Density 0.91
Quantity 5 g
Solubility Information Soluble in organic solvents.
Formula Weight 140.23
Percent Purity 99%
Chemical Name or Material Cyclohexyl ethyl ketone
EINECSNumber 214-380-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only