missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. A120825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A120825ML 25 mL
A1208500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A120825ML Supplier TCI America Supplier No. A120825ML

Chemical Identifiers

CAS 3066-71-5
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.21
MDL Number MFCD00046349
InChI Key KBLWLMPSVYBVDK-UHFFFAOYSA-N
Synonym cyclohexyl acrylate, 2-propenoic acid, cyclohexyl ester, sartomer sr 220, acrylic acid, cyclohexyl ester, acrylic acid cyclohexyl ester, cyclohexylacrylate, acrylic acid cyclohexyl, acmc-209slu, wln: l6tj aov1u1, 4-06-00-00038 beilstein handbook reference
PubChem CID 18298
IUPAC Name cyclohexyl prop-2-enoate
SMILES C=CC(=O)OC1CCCCC1

Specifications

Color Colorless
Boiling Point 184°C
Quantity 25 mL
UN Number 3082
Formula Weight 154.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl Acrylate (stabilized with MEHQ)
EINECSNumber (3)-2238
RTECSNumber AS7350000
TSCA Yes