Learn More
Cyclohexyl acrylate, 98+%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
$162.77 - $162.77
Chemical Identifiers
| CAS | 3066-71-5 |
|---|---|
| Molecular Formula | C9H14O2 |
| Molecular Weight (g/mol) | 154.21 |
| MDL Number | MFCD00046349 |
| InChI Key | KBLWLMPSVYBVDK-UHFFFAOYSA-N |
| Synonym | cyclohexyl acrylate, 2-propenoic acid, cyclohexyl ester, sartomer sr 220, acrylic acid, cyclohexyl ester, acrylic acid cyclohexyl ester, cyclohexylacrylate, acrylic acid cyclohexyl, acmc-209slu, wln: l6tj aov1u1, 4-06-00-00038 beilstein handbook reference |
| PubChem CID | 18298 |
| IUPAC Name | cyclohexyl prop-2-enoate |
| SMILES | C=CC(=O)OC1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1292514
|
Thermo Scientific Chemicals
L1292514 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3066-71-5 | |
| 154.21 | |
| KBLWLMPSVYBVDK-UHFFFAOYSA-N | |
| 18298 | |
| C=CC(=O)OC1CCCCC1 |
| C9H14O2 | |
| MFCD00046349 | |
| cyclohexyl acrylate, 2-propenoic acid, cyclohexyl ester, sartomer sr 220, acrylic acid, cyclohexyl ester, acrylic acid cyclohexyl ester, cyclohexylacrylate, acrylic acid cyclohexyl, acmc-209slu, wln: l6tj aov1u1, 4-06-00-00038 beilstein handbook reference | |
| cyclohexyl prop-2-enoate |
Specifications
| 3066-71-5 | |
| 182°C to 184°C | |
| C9H14O2 | |
| MFCD00046349 | |
| UN3082 | |
| cyclohexyl acrylate, 2-propenoic acid, cyclohexyl ester, sartomer sr 220, acrylic acid, cyclohexyl ester, acrylic acid cyclohexyl ester, cyclohexylacrylate, acrylic acid cyclohexyl, acmc-209slu, wln: l6tj aov1u1, 4-06-00-00038 beilstein handbook reference | |
| C=CC(=O)OC1CCCCC1 | |
| 154.21 | |
| 154.21 | |
| Cyclohexyl acrylate, stabilized with 100ppm 4-methoxyphenol |
| 0.98 | |
| 68°C (154°F) | |
| 1.459 | |
| 25 g | |
| 1937528 | |
| KBLWLMPSVYBVDK-UHFFFAOYSA-N | |
| cyclohexyl prop-2-enoate | |
| 18298 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H335-H227
Causes skin irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 221-319-3
RTECSNumber : AS7350000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only