missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Acetate 99.0+%, TCI America™

Catalog No. p-7116668
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A002825ML 25 mL
A0028500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A002825ML Supplier TCI America Supplier No. A002825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 622-45-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00003850
InChI Key YYLLIJHXUHJATK-UHFFFAOYSA-N
Synonym acetic acid, cyclohexyl ester, hexalin acetate, cyclohexanyl acetate, cyclohexanol, acetate, cyclohexanolazetat, adronal acetate, acetic acid cyclohexyl ester, cyclohexane acetate, cyclohexylester kyseliny octove, unii-ul0rs4h1ue
PubChem CID 12146
ChEBI CHEBI:31447
IUPAC Name cyclohexyl acetate
SMILES CC(=O)OC1CCCCC1

Specifications

Color Colorless
Boiling Point 174°C
Quantity 25 mL
UN Number 2243
Formula Weight 142.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl Acetate
EINECSNumber (3)-2316&(3)-2359
RTECSNumber AG5075000
TSCA Yes