CAS RN 622-45-7
IUPAC Name:
cyclohexyl acetate
Synonyms:
cyclohexyl acetate adronal acetate hexalin acetate cyclohexanolazetat cyclohexylester kyseliny octove
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
YYLLIJHXUHJATK-UHFFFAOYSA-N
SMILES:
CC(=O)OC1CCCCC1
1
–
1
of
1
results
1
Cyclohexyl Acetate 99.0+%, TCI America™
CAS: 622-45-7 Molecular Formula: C8H14O2 Molecular Weight (g/mol): 142.198 MDL Number: MFCD00003850 InChI Key: YYLLIJHXUHJATK-UHFFFAOYSA-N Synonym: acetic acid, cyclohexyl ester,hexalin acetate,cyclohexanyl acetate,cyclohexanol, acetate,cyclohexanolazetat,adronal acetate,acetic acid cyclohexyl ester,cyclohexane acetate,cyclohexylester kyseliny octove,unii-ul0rs4h1ue PubChem CID: 12146 ChEBI: CHEBI:31447 IUPAC Name: cyclohexyl acetate SMILES: CC(=O)OC1CCCCC1