missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene 99.0+%, TCI America™

Supplier:  TCI America C0491500ML

 View more versions of this product

Catalog No. C0491500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cyclohexene
-104°C
82°C
MFCD00001539
2256
HGCIXCUEYOPUTN-UHFFFAOYSA-N
cyclohexene
8079
82.15
Liquid
110-83-8
Colorless
C6H10
500 mL
tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
C1CCC=CC1
82.146
CHEBI:36404
≥99.0% (GC)

Chemical Identifiers

110-83-8
82.146
HGCIXCUEYOPUTN-UHFFFAOYSA-N
8079
cyclohexene
C6H10
MFCD00001539
tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
CHEBI:36404
C1CCC=CC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2234

RTECSNumber : GW2500000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions