missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene 99.0+%, TCI America™

Catalog No. p-7124464
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
C049125ML 25 mL
C0491500ML 500 mL
2 options

Catalog No. C049125ML

Supplier: TCI America C049125ML

Only null left

Chemical Identifiers

CAS 110-83-8
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00001539
InChI Key HGCIXCUEYOPUTN-UHFFFAOYSA-N
Synonym tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
PubChem CID 8079
ChEBI CHEBI:36404
IUPAC Name cyclohexene
SMILES C1CCC=CC1

Specifications

Melting Point -104°C
Color Colorless
Boiling Point 82°C
Quantity 25 mL
UN Number 2256
Formula Weight 82.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexene
EINECSNumber (3)-2234
RTECSNumber GW2500000
TSCA Yes