missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone Oxime 98.0+%, TCI America™

Catalog No. p-7124463
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
C049025G 25 g
C0490500G 500 g
2 options

Catalog No. C049025G

Supplier: TCI America C049025G

Only null left

Chemical Identifiers

CAS 100-64-1
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00001660
InChI Key VEZUQRBDRNJBJY-UHFFFAOYSA-N
Synonym cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
PubChem CID 7517
IUPAC Name N-cyclohexylidenehydroxylamine
SMILES ON=C1CCCCC1

Specifications

Melting Point 90°C
Boiling Point 210°C
Quantity 25 g
Formula Weight 113.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexanone Oxime
EINECSNumber (3)-2377
RTECSNumber GW1925000
TSCA Yes