accessibility menu, dialog, popup

UserName

CAS RN 100-64-1

IUPAC Name: N-cyclohexylidenehydroxylamine
Synonyms: cyclohexanone oxime
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: VEZUQRBDRNJBJY-UHFFFAOYSA-N
SMILES: ON=C1CCCCC1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)