missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone Oxime 98.0+%, TCI America™

Catalog No. C0490500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0490500G 500 g
C049025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0490500G Supplier TCI America Supplier No. C0490500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-64-1
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00001660
InChI Key VEZUQRBDRNJBJY-UHFFFAOYSA-N
Synonym cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
PubChem CID 7517
IUPAC Name N-cyclohexylidenehydroxylamine
SMILES ON=C1CCCCC1

Specifications

Melting Point 90°C
Boiling Point 210°C
Quantity 500 g
Formula Weight 113.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexanone Oxime
EINECSNumber (3)-2377
RTECSNumber GW1925000
TSCA Yes