missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone oxime, 97%

Catalog No. p-7024691
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1982022 100 g
AAA1982036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1982022 Supplier Thermo Scientific Chemicals Supplier No. A1982022
Only null left

CAS: 100-64-1 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-64-1
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00001660
InChI Key VEZUQRBDRNJBJY-UHFFFAOYSA-N
Synonym cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
PubChem CID 7517
IUPAC Name N-cyclohexylidenehydroxylamine
SMILES ON=C1CCCCC1

Specifications

Melting Point 87°C to 91°C
Boiling Point 206°C to 210°C
Flash Point 103°C (217°F)
Quantity 100 g
Beilstein 1616769
Formula Weight 113.16
Percent Purity 97%
Chemical Name or Material Cyclohexanone oxime
Hazard Category H228-H302-H319-H373
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P280-P301+P312-P305+P351+P338-P314-P330-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 202-874-0
RTECSNumber GW1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only