missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanol, 99%

Catalog No. AAA175760F
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA175760F 2500 mL
AAA17576AE 100 mL
AAA17576AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA175760F Supplier Thermo Scientific Chemicals Supplier No. A175760F
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 108-93-0 | C6H12O | 100.161 g/mol

Cyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.

Solubility
Miscible with ethanol, ethyl ether acetone, chloroform, ethyl acetate, linseed oil and benzene. Slightly miscible with cold water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, hydrogen peroxide and nitric acid.

Chemical Identifiers

CAS 108-93-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00003855
InChI Key HPXRVTGHNJAIIH-UHFFFAOYSA-N
Synonym cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol
PubChem CID 7966
ChEBI CHEBI:18099
IUPAC Name cyclohexanol
SMILES C1CCC(CC1)O

Specifications

Melting Point 23°C to 24°C
Density 0.951
Boiling Point 160°C to 161°C
Flash Point 67°C (152°F)
Assay Percent Range 99%
Refractive Index 1.465
Quantity 2500 mL
Beilstein 906744
Merck Index 14,2725
Solubility Information Miscible with ethanol,ethyl ether acetone,chloroform,ethyl acetate,linseed oil and benzene. Slightly miscible with cold water.
Formula Weight 100.16
Percent Purity 99%
Chemical Name or Material Cyclohexanol
Show More Show Less
Hazard Category H227-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H332-H315-H335-H227
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-630-6
RTECSNumber GV7875000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only