missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanol, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C146020LTMT

 View more versions of this product

Catalog No. 18610098


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cyclohexanol
1
20 L
100.16
Reagent
110-82-7
161.84°C
Steel Can
97%

Chemical Identifiers

110-82-7
100.16
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.