missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanol, 97%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 4 L |
| Packaging | Steel Can |
| Boiling Point | 161.84°C |
| Percent Purity | 97% |
| Grade | Reagent |
| Chemical Name or Material | Cyclohexanol |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.