missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanethiol 98.0+%, TCI America™
$113.71 - $602.10
Chemical Identifiers
| CAS | 1569-69-3 |
|---|---|
| Molecular Formula | C6H12S |
| Molecular Weight (g/mol) | 116.222 |
| MDL Number | MFCD00001455 |
| InChI Key | CMKBCTPCXZNQKX-UHFFFAOYSA-N |
| Synonym | cyclohexyl mercaptan, cyclohexyl thiol, cyclohexylmercaptan, cyclohexylthiol, cyklohexylmerkaptan, cyklohexanthiol, cyklohexanthiol czech, cyklohexylmerkaptan czech, hexahydrothiophenol, mercaptocyclohexane |
| PubChem CID | 15290 |
| IUPAC Name | cyclohexanethiol |
| SMILES | C1CCC(CC1)S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C072725ML
|
TCI America
C072725ML |
25 mL |
Each for $113.71
|
|
|||||
|
C0727500ML
|
TCI America
C0727500ML |
500 mL |
Each for $602.10
|
|
|||||
Chemical Identifiers
| 1569-69-3 | |
| 116.222 | |
| CMKBCTPCXZNQKX-UHFFFAOYSA-N | |
| 15290 | |
| C1CCC(CC1)S |
| C6H12S | |
| MFCD00001455 | |
| cyclohexyl mercaptan, cyclohexyl thiol, cyclohexylmercaptan, cyclohexylthiol, cyklohexylmerkaptan, cyklohexanthiol, cyklohexanthiol czech, cyklohexylmerkaptan czech, hexahydrothiophenol, mercaptocyclohexane | |
| cyclohexanethiol |
Specifications
| 1569-69-3 | |
| 158°C | |
| MFCD00001455 | |
| 3054 | |
| CMKBCTPCXZNQKX-UHFFFAOYSA-N | |
| cyclohexanethiol | |
| 15290 | |
| ≥98.0% (GC,T) | |
| Cyclohexanethiol |
| Colorless | |
| C6H12S | |
| 25 mL | |
| cyclohexyl mercaptan, cyclohexyl thiol, cyclohexylmercaptan, cyclohexylthiol, cyklohexylmerkaptan, cyklohexanthiol, cyklohexanthiol czech, cyklohexylmerkaptan czech, hexahydrothiophenol, mercaptocyclohexane | |
| C1CCC(CC1)S | |
| 116.222 | |
| 116.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2303
RTECSNumber : GV7525000
TSCA : Yes