missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanepropionic acid, 99%

Catalog No. p-96594
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171450500 50 g
AC171452500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC171450500 Supplier Thermo Scientific Chemicals Supplier No. 171450500

CAS: 701-97-3 | C9H16O2 | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 701-97-3
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD00001527
InChI Key HJZLEGIHUQOJBA-UHFFFAOYSA-N
Synonym cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid
PubChem CID 69702
IUPAC Name 3-cyclohexylpropanoic acid
SMILES OC(=O)CCC1CCCCC1

Specifications

Melting Point 14.0°C to 17.0°C
Color Colorless to Yellow
Density 0.9980g/mL
Boiling Point 275.7°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4630 to 1.4650
Linear Formula C6H11CH2CH2CO2H
Quantity 50 g
Beilstein 09, 82
Specific Gravity 0.998
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 156.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclohexanepropionic acid, 99%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes serious eye irritation. Harmful if swallowed. Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 211-861-9
TSCA TSCA
Recommended Storage Product may darken during storage

RUO – Research Use Only