missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexaneethanol 98.0+%, TCI America™

Catalog No. p-7124511
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C058225ML 25 mL
C0582500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C058225ML Supplier TCI America Supplier No. C058225ML
Only null left

Chemical Identifiers

CAS 4442-79-9
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001525
InChI Key QJQZRLXDLORINA-UHFFFAOYSA-N
Synonym cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol
PubChem CID 20508
IUPAC Name 2-cyclohexylethanol
SMILES C1CCC(CC1)CCO

Specifications

Color Colorless
Boiling Point 206°C
Quantity 25 mL
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexaneethanol
EINECSNumber (3)-2307
RTECSNumber KK3528000
TSCA Yes