accessibility menu, dialog, popup

UserName

OH

CAS RN 4442-79-9

IUPAC Name: 2-cyclohexylethan-1-ol
Synonyms: cyclohexylethanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: QJQZRLXDLORINA-UHFFFAOYSA-N
SMILES: OCCC1CCCCC1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)