missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanecarbonyl Chloride 98.0+%, TCI America™
Supplier: TCI America C068525ML
Specifications
| Cyclohexanecarbonyl Chloride | |
| Yellow | |
| C7H11ClO | |
| 25 mL | |
| hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride | |
| ClC(=O)C1CCCCC1 | |
| 146.61 | |
| 146.61 | |
| Liquid |
| 2719-27-9 | |
| 185°C | |
| MFCD00001456 | |
| 3265 | |
| RVOJTCZRIKWHDX-UHFFFAOYSA-N | |
| cyclohexanecarbonyl chloride | |
| 75938 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2719-27-9 | |
| 146.61 | |
| RVOJTCZRIKWHDX-UHFFFAOYSA-N | |
| 75938 | |
| ClC(=O)C1CCCCC1 |
| C7H11ClO | |
| MFCD00001456 | |
| hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride | |
| cyclohexanecarbonyl chloride |
Safety and Handling
EINECSNumber : (3)-2354
TSCA : Yes