missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarbonyl Chloride 98.0+%, TCI America™

Catalog No. p-7124561
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C068525ML 25 mL
C0685100ML 100 mL
C0685500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C068525ML Supplier TCI America Supplier No. C068525ML
Only null left

Chemical Identifiers

CAS 2719-27-9
Molecular Formula C7H11ClO
Molecular Weight (g/mol) 146.61
MDL Number MFCD00001456
InChI Key RVOJTCZRIKWHDX-UHFFFAOYSA-N
Synonym hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride
PubChem CID 75938
IUPAC Name cyclohexanecarbonyl chloride
SMILES ClC(=O)C1CCCCC1

Specifications

Color Yellow
Boiling Point 185°C
Quantity 25 mL
UN Number 3265
Formula Weight 146.61
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexanecarbonyl Chloride
EINECSNumber (3)-2354
TSCA Yes