missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanone Oxime 98.0+%, TCI America™

Catalog No. C154325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C154325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C154325G Supplier TCI America Supplier No. C154325G
Only null left

Chemical Identifiers

CAS 2158-31-8
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00046508
InChI Key OENGSNXUALAIFP-UHFFFAOYSA-N
Synonym Suberoxime
PubChem CID 137457
IUPAC Name N-cycloheptylidenehydroxylamine
SMILES C1CCCC(=NO)CC1

Specifications

Color Yellow
Boiling Point 152°C
Quantity 25 g
Formula Weight 127.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cycloheptanone Oxime