accessibility menu, dialog, popup

UserName

HON

CAS RN 2158-31-8

IUPAC Name: N-cycloheptylidenehydroxylamine
Synonyms: N-cycloheptylidenehydroxylamine
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: OENGSNXUALAIFP-UHFFFAOYSA-N
SMILES: ON=C1CCCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results