missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanecarboxylic acid, 95%

Catalog No. AC111020050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111020050 5 g Glass bottle
AC111020250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111020050 Supplier Thermo Scientific Chemicals Supplier No. 111020050

CAS: 1460-16-8 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1460-16-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00004152
InChI Key VZFUCHSFHOYXIS-UHFFFAOYSA-N
Synonym cycloheptane carboxylic acid, cycloheptanoic acid, cycloheptanecarboxylicacid, cyclohepatan carboxylic acid, acmc-1c1rc, sampl4, o9, vzfuchsfhoyxis-uhfffaoysa, cycloheptanecarboxylic acid
PubChem CID 15091
ChEBI CHEBI:29564
IUPAC Name cycloheptanecarboxylic acid
SMILES OC(=O)C1CCCCCC1

Specifications

Color Colorless
Density 1.0350g/mL
Boiling Point 135.0°C to 138.0°C (9.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4710 to 1.4730 (20°C, 589nm)
Linear Formula C7H13CO2H
Quantity 5 g
Beilstein 09, 12
Specific Gravity 1.035
Formula Weight 142.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cycloheptanecarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-954-5

RUO – Research Use Only