CAS RN 1460-16-8
IUPAC Name:
cycloheptanecarboxylic acid
Synonyms:
cycloheptanecarboxylic acid
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
VZFUCHSFHOYXIS-UHFFFAOYSA-N
SMILES:
OC(=O)C1CCCCCC1
1
–
1
of
1
results
1
Promotions Available
Cycloheptanecarboxylic acid, 95%
Cycloheptanecarboxylic acid, 95%
CAS: 1460-16-8 Molecular Formula: C8H14O2 Molecular Weight (g/mol): 142.20 MDL Number: MFCD00004152 InChI Key: VZFUCHSFHOYXIS-UHFFFAOYSA-N Synonym: cycloheptane carboxylic acid,cycloheptanoic acid,cycloheptanecarboxylicacid,cyclohepatan carboxylic acid,acmc-1c1rc,sampl4, o9,vzfuchsfhoyxis-uhfffaoysa,cycloheptanecarboxylic acid PubChem CID: 15091 ChEBI: CHEBI:29564 IUPAC Name: cycloheptanecarboxylic acid SMILES: OC(=O)C1CCCCCC1