missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclodextrin hydrate, 98+%

Catalog No. AC227290050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227290050 5 g Glass bottle
AC227290250 25 g Glass bottle
AC227291000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227290050 Supplier Thermo Scientific Chemicals Supplier No. 227290050

CAS: 51211-51-9 | C36H60O30 | 972.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 51211-51-9
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.85
MDL Number MFCD00150804
InChI Key HFHDHCJBZVLPGP-UHFFFAOYNA-N
Synonym alpha-cyclodextrin hydrate, water.beta-cyclodextrin, cyclohexaamylose hydrate, ^a-cyclodextrin hydrate, alpha-cyclodextrin water, 972.86 anhydrous
PubChem CID 60161466
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 278°C
Color White
Loss on Drying 11% max.
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. Linear oligomers (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 14, 2718
Specific Rotation + 148
Solubility Information Solubility in water: 145g/L (25°C)
IUPAC Name 5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
Formula Weight 972.85
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material α-Cyclodextrin hydrate
Show More Show Less

RUO – Research Use Only