CAS RN 51211-51-9
IUPAC Name:
5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
Synonyms:
α-CD|α-dextrin|α-cycloamylose|cyclohexaamylose|cyclomaltohexaose|α-schardinger dextrin|cyclohexapentylose|cyclomaltohexose|alfadexum|α-cyclo dextrin
Molecular Weight (g/mol):
972.85
Molecular Formula:
C36H60O30
InChi Key:
HFHDHCJBZVLPGP-UHFFFAOYNA-N
SMILES:
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
1
–
1
of
1
results
1
Promotions Available
alpha-Cyclodextrin hydrate, 98+%
alpha-Cyclodextrin hydrate, 98+%
CAS: 51211-51-9 Molecular Formula: C36H60O30 Molecular Weight (g/mol): 972.85 MDL Number: MFCD00150804 InChI Key: HFHDHCJBZVLPGP-UHFFFAOYNA-N Synonym: alpha-cyclodextrin hydrate,water.beta-cyclodextrin,cyclohexaamylose hydrate,^a-cyclodextrin hydrate,alpha-cyclodextrin water,972.86 anhydrous PubChem CID: 60161466 SMILES: OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O