missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutylamine hydrochloride, 97%

Catalog No. p-7028215
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2540006 5 g
AAB2540014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2540006 Supplier Thermo Scientific Chemicals Supplier No. B2540006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6291-01-6 | C4H10ClN | 107.58 g/mol

Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool, dry conditions, in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6291-01-6
Molecular Formula C4H10ClN
Molecular Weight (g/mol) 107.58
MDL Number MFCD00034953
InChI Key NFAZOGXQOWEWBM-UHFFFAOYSA-N
Synonym cyclobutanamine hydrochloride, cyclobutylamine hydrochloride, aminocyclobutane hcl, cyclobutanamine, hydrochloride, cyclobutanaminium chloride, cyclobutylamine hcl, acmc-209wdt, aminocyclobutane hydrochloride, cyclobutylamine hydrochloric acid, cyclobutylamine hydrochloride salt
PubChem CID 12618374
IUPAC Name cyclobutanamine;hydrochloride
SMILES C1CC(C1)N.Cl

Specifications

Melting Point 180°C to 183°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 107.58
Percent Purity 97%
Chemical Name or Material Cyclobutylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-540-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only