CAS RN 6291-01-6
IUPAC Name:
hydrogen cyclobutanamine chloride
Synonyms:
hydrogen cyclobutylamine chloride
Molecular Weight (g/mol):
107.58
Molecular Formula:
C4H10ClN
InChi Key:
NFAZOGXQOWEWBM-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NC1CCC1
1
–
1
of
1
results
1
Promotions Available
Cyclobutylamine hydrochloride, 97%
Cyclobutylamine hydrochloride, 97%
CAS: 6291-01-6 Molecular Formula: C4H10ClN Molecular Weight (g/mol): 107.58 MDL Number: MFCD00034953 InChI Key: NFAZOGXQOWEWBM-UHFFFAOYSA-N Synonym: cyclobutanamine hydrochloride,cyclobutylamine hydrochloride,aminocyclobutane hcl,cyclobutanamine, hydrochloride,cyclobutanaminium chloride,cyclobutylamine hcl,acmc-209wdt,aminocyclobutane hydrochloride,cyclobutylamine hydrochloric acid,cyclobutylamine hydrochloride salt PubChem CID: 12618374 IUPAC Name: cyclobutanamine;hydrochloride SMILES: C1CC(C1)N.Cl