missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutyl methyl ketone, 97%

CAS: 3019-25-8 | C6H10O | 98.145 g/mol

Supplier:  Thermo Scientific Chemicals L0352106

 View more versions of this product

Catalog No. AAL0352106


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclobutyl methyl ketone
0.9
28°C (82°F)
1.432
5 g
1924022
JPJOOTWNILDNAW-UHFFFAOYSA-N
1-cyclobutylethanone
76398
97%
3019-25-8
134°C to 135°C
C6H10O
MFCD00037148
UN1224
cyclobutyl methyl ketone, 1-cyclobutylethan-1-one, acetylcyclobutane, ethanone, 1-cyclobutyl, ketone, cyclobutyl methyl, methyl cyclobutyl ketone, cyclobutylmethylketone, acetylcyclobutan, cyclobutylethanone, cyclobutane, acetyl
CC(=O)C1CCC1
98.145
98.15

Chemical Identifiers

3019-25-8
98.145
JPJOOTWNILDNAW-UHFFFAOYSA-N
76398
CC(=O)C1CCC1
C6H10O
MFCD00037148
cyclobutyl methyl ketone, 1-cyclobutylethan-1-one, acetylcyclobutane, ethanone, 1-cyclobutyl, ketone, cyclobutyl methyl, methyl cyclobutyl ketone, cyclobutylmethylketone, acetylcyclobutan, cyclobutylethanone, cyclobutane, acetyl
1-cyclobutylethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c

H226-H302

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 221-163-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only