missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutyl methyl ketone, 97%

Catalog No. p-7043930
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0352103 1 g
AAL0352106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0352103 Supplier Thermo Scientific Chemicals Supplier No. L0352103
Only null left

CAS: 3019-25-8 | C6H10O | 98.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3019-25-8
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00037148
InChI Key JPJOOTWNILDNAW-UHFFFAOYSA-N
Synonym cyclobutyl methyl ketone, 1-cyclobutylethan-1-one, acetylcyclobutane, ethanone, 1-cyclobutyl, ketone, cyclobutyl methyl, methyl cyclobutyl ketone, cyclobutylmethylketone, acetylcyclobutan, cyclobutylethanone, cyclobutane, acetyl
PubChem CID 76398
IUPAC Name 1-cyclobutylethanone
SMILES CC(=O)C1CCC1

Specifications

Density 0.9
Boiling Point 134°C to 135°C
Flash Point 28°C (82°F)
Refractive Index 1.432
Quantity 1 g
UN Number UN1224
Beilstein 1924022
Formula Weight 98.15
Percent Purity 97%
Chemical Name or Material Cyclobutyl methyl ketone
Hazard Category H226-H302
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 221-163-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only