missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanemethanol 98.0+%, TCI America™

Catalog No. C20615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20615G 5 g
C206125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20615G Supplier TCI America Supplier No. C20615G

Chemical Identifiers

CAS 4415-82-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00001330
InChI Key WPOPOPFNZYPKAV-UHFFFAOYSA-N
Synonym cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465
PubChem CID 78130
IUPAC Name cyclobutylmethanol
SMILES C1CC(C1)CO

Specifications

Color Colorless
Boiling Point 144°C
Quantity 5 g
UN Number 1987
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclobutanemethanol