missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanemethanol, 99%, Thermo Scientific™

Chemical Identifiers

CAS 4415-82-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00001330
InChI Key WPOPOPFNZYPKAV-UHFFFAOYSA-N
Synonym cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465
PubChem CID 78130
IUPAC Name cyclobutylmethanol
SMILES C1CC(C1)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC182230050
SDS View Documents
Thermo Scientific Chemicals
182230050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC182230250
SDS View Documents
Thermo Scientific Chemicals
182230250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4415-82-1
86.13
WPOPOPFNZYPKAV-UHFFFAOYSA-N
78130
C1CC(C1)CO
C5H10O
MFCD00001330
cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465
cyclobutylmethanol

Specifications

4415-82-1
143°C to 144°C
Authentic
Glass bottle
1.4450 to 1.447
MFCD00001330
0.91
Solubility in water: immiscible
C1CC(C1)CO
86.13
86.13
Cyclobutanemethanol, 99%
0.9100g/mL
40°C
99%
C5H10O
C4H7CH2OH
5g
cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465
WPOPOPFNZYPKAV-UHFFFAOYSA-N
cyclobutylmethanol
78130
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-575-4

SDS
Documents

Documents

RUO – Research Use Only