Learn More
Cyclobutanemethanol, 99%
CAS: 4415-82-1 | C5H10O | 86.134 g/mol
$222.42 - $222.42
Chemical Identifiers
| CAS | 4415-82-1 |
|---|---|
| Molecular Formula | C5H10O |
| Molecular Weight (g/mol) | 86.134 |
| MDL Number | MFCD00001330 |
| InChI Key | WPOPOPFNZYPKAV-UHFFFAOYSA-N |
| Synonym | cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 |
| PubChem CID | 78130 |
| IUPAC Name | cyclobutylmethanol |
| SMILES | C1CC(C1)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2148014
|
Thermo Scientific Chemicals
B2148014 |
25 g |
Each for $222.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4415-82-1 | |
| 86.134 | |
| WPOPOPFNZYPKAV-UHFFFAOYSA-N | |
| 78130 | |
| C1CC(C1)CO |
| C5H10O | |
| MFCD00001330 | |
| cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 | |
| cyclobutylmethanol |
Specifications
| 4415-82-1 | |
| 143°C to 144°C | |
| C5H10O | |
| MFCD00001330 | |
| UN1105 | |
| cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 | |
| C1CC(C1)CO | |
| 86.134 | |
| 86.13 | |
| Cyclobutanemethanol |
| 0.913 | |
| 39°C (102°F) | |
| 1.4465 | |
| 25 g | |
| 2036027 | |
| WPOPOPFNZYPKAV-UHFFFAOYSA-N | |
| cyclobutylmethanol | |
| 78130 | |
| 99% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
EINECSNumber : 224-575-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only