Learn More
Cyclobutanecarbonyl chloride, 98%
CAS: 5006-22-4 | C5H7ClO | 118.56 g/mol
$197.71 - $640.81
Chemical Identifiers
| CAS | 5006-22-4 |
|---|---|
| Molecular Formula | C5H7ClO |
| Molecular Weight (g/mol) | 118.56 |
| MDL Number | MFCD00001319 |
| InChI Key | JFWMYCVMQSLLOO-UHFFFAOYSA-N |
| Synonym | cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1 |
| PubChem CID | 78705 |
| IUPAC Name | cyclobutanecarbonyl chloride |
| SMILES | C1CC(C1)C(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2031709
|
Thermo Scientific Chemicals
B2031709 |
10 g |
N/A
|
|
|||||
|
AAB2031718
|
Thermo Scientific Chemicals
B2031718 |
50 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5006-22-4 | |
| 118.56 | |
| JFWMYCVMQSLLOO-UHFFFAOYSA-N | |
| 78705 | |
| C1CC(C1)C(=O)Cl |
| C5H7ClO | |
| MFCD00001319 | |
| cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1 | |
| cyclobutanecarbonyl chloride |
Specifications
| 5006-22-4 | |
| 145°C | |
| Stench | |
| 1.455 | |
| 10 g | |
| 956624 | |
| cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1 | |
| C1CC(C1)C(=O)Cl | |
| 118.56 | |
| 118.56 | |
| Cyclobutanecarbonyl chloride |
| 1.039 | |
| 37°C (99°F) | |
| C5H7ClO | |
| MFCD00001319 | |
| UN2920 | |
| Moisture Sensitive | |
| JFWMYCVMQSLLOO-UHFFFAOYSA-N | |
| cyclobutanecarbonyl chloride | |
| 78705 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 225-680-8
TSCA : No
RUO – Research Use Only