missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarbonyl chloride, 98%

CAS: 5006-22-4 | C5H7ClO | 118.56 g/mol

Supplier:  Thermo Scientific Chemicals B2031718

 View more versions of this product

Catalog No. AAB2031718


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclobutanecarbonyl chloride
1.039
37°C (99°F)
C5H7ClO
MFCD00001319
UN2920
Moisture Sensitive
JFWMYCVMQSLLOO-UHFFFAOYSA-N
cyclobutanecarbonyl chloride
78705
98%
5006-22-4
145°C
Stench
1.455
50 g
956624
cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
C1CC(C1)C(=O)Cl
118.56
118.56

Chemical Identifiers

5006-22-4
118.56
JFWMYCVMQSLLOO-UHFFFAOYSA-N
78705
C1CC(C1)C(=O)Cl
C5H7ClO
MFCD00001319
cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
cyclobutanecarbonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

EINECSNumber : 225-680-8

TSCA : No

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only