missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarbonitrile, 97%

Catalog No. AAA1030914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1030914 25 g
AAA1030906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1030914 Supplier Thermo Scientific Chemicals Supplier No. A1030914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4426-11-3 | C5H7N | 81.12 g/mol

It is used for research or analysis use only, as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for research or analysis use only, as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, acids.

Chemical Identifiers

CAS 4426-11-3
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.12
MDL Number MFCD00013735
InChI Key GFBLFDSCAKHHGX-UHFFFAOYSA-N

Specifications

Density 0.868
Boiling Point 144°C to 146°C
Flash Point 42°C (107°F)
Odor Recognizable
Refractive Index 1.4325
Quantity 25 g
UN Number UN1992
Beilstein 2037573
Synonym cyanocyclobutane, cyclobutyl cyanide, cyclobutancarbonitrile, cyclobutane carbonitrile, acmc-1afpy, cyclobutanecarbonitrile 5g, cyanocyclobutane; cyclobutyl cyanide
Solubility Information Slightly soluble in water.
SMILES C1CC(C1)C#N
IUPAC Name cyclobutanecarbonitrile
PubChem CID 78148
Formula Weight 81.12
Percent Purity 97%
Chemical Name or Material Cyclobutanecarbonitrile
Show More Show Less
Hazard Category H226-H302+H312+H332
Hazard Statement GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 224-606-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only