accessibility menu, dialog, popup

UserName

N

CAS RN 4426-11-3

IUPAC Name: cyclobutanecarbonitrile
Synonyms: cyclobutanecarbonitrile
Molecular Weight (g/mol): 81.12
Molecular Formula: C5H7N
InChi Key: GFBLFDSCAKHHGX-UHFFFAOYSA-N
SMILES: N#CC1CCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)