missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Cyclobenzaprine Hydrochloride, USP
Description
Cyclobenzaprine Hydrochloride USP is a white to off-white, odorless, crystalline powder. It is freely soluble in water, in alcohol, and in methanol.
Specifications
Specifications
| Chemical Name or Material | Cyclobenzaprine Hydrochloride, USP |
| Packaging | Jar 1500ml, White, HDPE, Round, 89-400, TRIVENI-DB, CT |
| CAS | 6202-23-9 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 311.85 |
| Quantity | 500 g |
| Percent Purity | 98 to 102 % |
| Grade | USP |